missing translation for 'onlineSavingsMsg'
Få mere at vide

Thermo Scientific Chemicals Crystal Violet

Artikelnummer. 11455027
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
25 g
100 g
500 g
Pakningsstørrelse:
100g
25g
500g
Artikelnummer. Quantity unitSize
11455027 500 g 500g
11435027 25 g 25g
11445027 100 g 100g
3 options
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 11455027

Brand: Thermo Scientific Chemicals B21932.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol

Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.

Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
MDL Number MFCD00011750
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Basic Violet 3; C.I. 42555
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Tekniske data

Melting Point ∼215°C (decomposition)
UN Number UN3077
Beilstein 4077708
Merck Index 14,4395
Solubility Information Miscible with water and chloroform. Partially miscible with alcohol and glycerol.
Formula Weight 407.99
Quantity 500 g
Chemical Name or Material Crystal Violet

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.