missing translation for 'onlineSavingsMsg'
Learn More

Crystal Ponceau 6R (Technical Grade), TRC

Product Code. 30357468
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357468 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357468 Supplier Toronto Research Chemicals Supplier No. TRCC8274901G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acid Red 44
CAS 2766-77-0
Molecular Formula C20 H12 N2 O7 S2 . 2 Na
Quantity 1 g
Synonym 1,3-Naphthalenedisulfonic acid, 7-hydroxy-8-(1-naphthalenylazo)-, disodium salt (9CI), 2-Naphthol-6,8-disulfonic acid, 1-(1-naphthylazo)-, disodium salt (6CI), C.I. Acid Red 44 (7CI), C.I. Acid Red 44, disodium salt (8CI), Acid Leather Ponceau 6R, Acid Ponceau 6R, Acid Red 6A, Acidal Crystals Ponceau, Brilliant Crystal Scarlet, C.I. 16250, Colacid Red 6A, Crystal Ponceau, Crystal Ponceau 6R, Crystal Ponceau M 6R, Crystal Scarlet, Crystal Scarlet 6R, Crystal Tertracid Ponceau 6R, Kemacid Leather Ponceau 6R, Ponceau 6R, Ponceau 6R Crystals, Ponceau Cristallise Extra A Export, Ponceau Crystals 6R, Romexal Red 6A, Triacid Ponceau 6R
InChI Formula InChI=1S/C20H14N2O7S2.2Na/c23-17-9-8-13-10-14(30(24,25)26)11-18(31(27,28)29)19(13)20(17)22-21-16-7-3-5-12-4-1-2-6-15(12)16;;/h1-11,23H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2
SMILES [Na+].[Na+].Oc1ccc2cc(cc(c2c1N=Nc3cccc4ccccc34)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name disodium;7-hydroxy-8-[(E)-naphthalen-1-yldiazenyl]naphthalene-1,3-disulfonate
Molecular Weight (g/mol) 502.43
Formula Weight 501.9881
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.