missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ LMT-55 Tubing without Configuration

Product Code. 15183226
Change view
Click to view available options
Diameter (Metric) Inner:
0.51 mm
0.64 mm
0.89 mm
0.95 mm
1.02 mm
1.14 mm
1.65 mm
2.06 mm
2.79 mm
Color:
Blue/Blue
Crystal Clear
Orange/White
Orange/Yellow
Purple/Purple
Purple/White
Red/Red
White/Black
Unit Size:
10m
Product Code. Diameter (Metric) Inner Color unitSize
15183226 1.14 mm Red/Red 10m
15113226 0.64 mm Orange/White 10m
15153226 0.95 mm White/Black 10m
10092393 1.65 mm Blue/Blue 10m
15183216 0.51 mm Orange/Yellow 10m
15123226 0.89 mm Crystal Clear 10m
15133236 2.06 mm Purple/Purple 10m
15133226 1.02 mm Crystal Clear 10m
12068908 2.79 mm Purple/White 10m
9 options
This item is not returnable. View return policy

Product Code. 15183226

Brand: Saint Gobain Life Sciences 07053413LND

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing without Configuration handles virtually all inorganic chemicals found in the lab.

  • Bio-based formulation delivers excellent performance
  • Lot-to-lot consistency for reproducible results
  • Non-toxic and non-contaminating
  • Smooth, polished inner wall
  • Slips easily over fittings and grips securely for simple lab set-ups
  • Contains no BPA or phthalates
  • Standard sizes available to hold full vacuum at room temperature
Typical Applications
  • General laboratory
  • Food and beverage
  • Biopharmaceutical
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain line
  • Resistant to flex fatigue and abrasion
  • Less permeable than rubber tubing
  • For use with virtually all inorganic chemicals found in the lab
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP Class VI
Color Code Red/Red
Description 1.14 x 0.85 mm
Diameter (English) Inner 0.04 in.
Diameter (Metric) Inner 1.14 mm
Material Tygon R3607
Overall Length (English) 32.8 ft.
Overall Length (Metric) 10 m
Thickness (English) 0.03 in.
Thickness (Metric) 0.85 mm
For Use With (Application) For sample and reagent dispensing, biological and chemical assays, and water and soil analysis
Min. Operating Temperature (English) -58°F
Min. Operating Temperature (Metric) -50°C
Packaging Quantity 1 Ea.
Color Red/Red
Max. Operating Temperature (English) 165.2°F
Max. Operating Temperature (Metric) 74°C
Product Type Tubing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.