Learn More
Saint Gobain Life Sciences Tygon™ E-3603 Non-DEHP Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ E-3603 Non-DEHP Tubing handles virtually all inorganic chemicals found in the lab.
Brand: Saint Gobain Life Sciences ACF00014-C
Code : 02
Additional Details : Weight : 1.26400kg
Description
- The new bio-based formulation delivers the same excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Tubing | |
0.19 in. | |
0.44 in. | |
49.2 ft. | |
1/8 in. | |
Crystal Clear |
FDA Compliant, NSF, 3-A, USP Class VI, Japan Food Sanitation Law (#370/1959,1935/2004/EC and 10/2011/EU) | |
4.8 mm | |
11.20 mm | |
15 m | |
3.2 mm | |
74°C |