missing translation for 'onlineSavingsMsg'
Learn More

Cryptotanshinone, TRC

Product Code. 30356478
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
50 mg
500 mg
missing translation for 'unitSize':
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30356478 25 mg 25mg
30365240 500 mg -
30355740 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30356478 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC82748025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35825-57-1
Molecular Formula C19 H20 O3
Molecular Weight (g/mol) 296.36
Synonym Phenanthro[1,2-b]furan-10,11-dione, 1,2,6,7,8,9-hexahydro-1,6,6-trimethyl-, (1R)-, Phenanthro[1,2-b]furan-10,11-dione, 1,2,6,7,8,9-hexahydro-1,6,6-trimethyl-, (R)- (8CI), (15R)-Cryptotanshinone, (1R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione, Cryototanshinone, Cryptotanshinon, Cryptotanshinone, Tanshinone c
IUPAC Name (1R)-1,6,6-trimethyl-2,7,8,9-tetrahydro-1H-naphtho[1,2-g][1]benzofuran-10,11-dione
SMILES C[C@H]1COC2=C1C(=O)C(=O)c3c4CCCC(C)(C)c4ccc23

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H20O3/c1-10-9-22-18-12-6-7-13-11(5-4-8-19(13,2)3)15(12)17(21)16(20)14(10)18/h6-7,10H,4-5,8-9H2,1-3H3/t10-/m0/s1
Formula Weight 296.1412
Chemical Name or Material Crypto-Tanshinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.