missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cryptosporidium parvum Oocysts Monoclonal Antibody (BEL 0126)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183095
This item is not returnable.
View return policy
Description
Mouse anti Cryptosporidium antibody, clone BEL 0126 recognizes a membrane antigen expressed by the oocysts of Cryptosporidium sp., an obligate enteric coccidian parasite of the phylum Apicomplexa that infects the gastrointestinal tract.
Specifications
| Cryptosporidium parvum Oocysts | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Protozoa | |
| Antibody | |
| IgG3 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| BEL 0126 | |
| Unconjugated | |
| Cryptosporidiosis | |
| Purified oocysts from bovine faeces. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction