missing translation for 'onlineSavingsMsg'
Learn More

Cryptochlorogenic acid, MedChemExpress

Product Code. 16461508
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461508 5 mg 5mg
16489098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461508 Supplier Medchem Express Supplier No. HYN0787
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cryptochlorogenic acid is a natural product.

Cryptochlorogenic acid is a natural product.

Specifications

Chemical Name or Material Cryptochlorogenic acid
CAS 905-99-7
Color White
Purity Grade Notes Research
Molecular Formula C16H18O9
Quantity 5 mg
Synonym 4-Caffeoylquinic acid 4-O-Caffeoylquinic acid
Solubility Information DMSO : 50 mg/mL (141.12 mM; Need ultrasonic)
SMILES O=C(/C=C/C1=CC(O)=C(O)C=C1)O[C@@H]([C@@H]2O)[C@@H](C[C@](C(O)=O)(O)C2)O
Molecular Weight (g/mol) 354.31
Formula Weight 354.31
Percent Purity 99.07%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.