Learn More
Thermo Scientific™ Cryotome™ Cryostat Accessories, Camel hair brush
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
Brand: Thermo Scientific™ 1910
This item is not returnable.
View return policy
Description
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
- Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
- Freezing time is shortened if freezing stage and tissue are placed on active Peltier
- Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples