missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CryoStor™ CSB
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
500mL
Description
- CryoStor™ CSB is an intracellular-like base formulation from the CryoStor™ preservation media series
- It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO)
- It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death
Specifications
Specifications
| Cell Type | B Cells, CHO Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells |
| For Use With (Application) | Cryopreservation |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction