missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CryoStor™ CSB

Product Code. 17157501
Encompass_Preferred
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
500mL
This item is not returnable. View return policy

Product Code. 17157501

Brand: STEMCELL Technologies 1000239

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Animal component-free and protein-free cryopreservation formulation

  • CryoStor™ CSB is an intracellular-like base formulation from the CryoStor™ preservation media series
  • It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO)
  • It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death

Specifications

Cell Type B Cells, CHO Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells
For Use With (Application) Cryopreservation
Quantity 1000 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.