missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CryoStor™ CS5

Product Code. p-7217827
Encompass_Preferred
Click to view available options
Quantity:
100 mL
100 mL Bag
5 x 10 mL
Unit Size:
100mL
Pack of 5
This item is not returnable. View return policy

Product Code. 17119931

Brand: STEMCELL Technologies 07949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Animal-component-free, defined cryopreservation medium with 5% DMSO

  • CryoStor™ CS5 is a uniquely formulated serum-free, animal component-free and defined cryopreservation medium containing 5% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS5 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS5 is cGMP-manufactured with USP grade components

Specifications

Cell Type CHO Cells, Mesenchymal Stem and Progenitor Cells, Other, Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 5 x 10 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.