missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CryoStor™ CS5

Product Code. 17129921
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
100 mL Bag
5 x 10 mL
Unit Size:
100mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17129921 100 mL 100mL
17119931 5 x 10 mL Pack of 5
17149931 100 mL Bag 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17129921 Supplier STEMCELL Technologies Supplier No. 07933
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Animal-component-free, defined cryopreservation medium with 5% DMSO

  • CryoStor™ CS5 is a uniquely formulated serum-free, animal component-free and defined cryopreservation medium containing 5% dimethyl sulfoxide (DMSO)
  • Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS5 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
  • CryoStor™ CS5 is cGMP-manufactured with USP grade components

Specifications

Cell Type CHO Cells, Mesenchymal Stem and Progenitor Cells, Other, Pluripotent Stem Cells
For Use With (Application) Cryopreservation
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.