missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CryoStor™ CS10
Descrizione
- CryoStor™ CS10 is a uniquely formulated serum-free, animal component-free, and defined cryopreservation medium containing 10% dimethyl sulfoxide (DMSO)
- Designed to preserve cells in low temperature environments (-80°C to -196°C), CryoStor™ CS10 provides a safe, protective environment for cells and tissues during the freezing and thawing processes and during storage
- CryoStor™ CS10 is recommended for the cryopreservation of hepatocytes, tissue samples, human peripheral blood, CHO cells, myeloma cell lines, hybridomas, human mesenchymal stem cells, human embryonic and human induced pluripotent stem cells (ES cells and iPS cells) and other extremely sensitive cell types
- CryoStor™ CS10 is cGMP-manufactured with USP grade components
Specifica
Specifica
| Cell Type | B Cells, CHO Cells, Hematopoietic Stem and Progenitor Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells |
| For Use With (Application) | Cryopreservation |
| Quantity | 1000 mL Bag |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?