Learn More
Invitrogen™ DEPDC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113948
Description
Immunogen sequence: EAVPTKVFGK DKKPTFEDSS CSLYRFTTIP NQDSQLGKEN KLYSPARYAD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 86%; Rat: 93%.
DEPDC7 gene ontology annotations related to this gene include intracellular signal transduction; regulation of small GTPase mediated signal transduction.
Specifications
| DEPDC7 | |
| Polyclonal | |
| Unconjugated | |
| DEPDC7 | |
| AV216087; DEP domain containing 7; DEP domain-containing protein 7; Depdc7; dJ85M6.4; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15; RGD1309720; TR2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 91614 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96QD5 | |
| DEPDC7 | |
| Synthetic peptide directed towards the N terminal region of human DEPDC7 (aa 109-158). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.