missing translation for 'onlineSavingsMsg'
Learn More

Shamrock™ Cryogenic Labels in Rolls for Thermal Printers

Product Code. 11802661
Click to view available options
Size (Metric):
12.7 x 23mm
12.7 x 38 mm
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11802661

Brand: Shamrock™ FCS515R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These labels are for use with all cryogenic temperatures, provided on rolls of 1000 labels.

  • Glove friendly
  • Smudge resistant
  • Available in two sizes
  • 1000 labels per roll

Specifications

Color White
For Use With (Application) Cryogenic temperatures
Quantity 1000/Pk.
Size (Metric) 12.7 x 38 mm
Dimensions (LxW) 38 x 12.7 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.