missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Cryogenic Gloves
Description
- Water-repellent silicone cowhide grain leather glove
- Fully lined with 2 aluminum/fleece layers
- Gunn cut, winged thumb, crispin type
- Reinforcement between thumb and index
- 200 mm split leather cuff
- Velcro™ adjustment strap on side of the palm
- Assembly, the winged thumb and flexibility of the materials used make this glove anatomical, thus providing excellent comfort
- The Velcro™ fastening tab gives the glove good support
- The silicone cowhide grain leather ensures good flexibility, even at a low temperature as well as water repellence and good mechanical protection
- Its polar double insulation offers outstanding protection against the cold and rays
- The reinforcement between the thumb and index finger increases the glove's mechanical resistance and durability by strengthening the areas of wear
- The hide cuff offers good forearm protection against mechanical hazards and limits any risk of intrusion
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical Hazards, EN 511-2006 Cold protection |
| Color | Yellow, White |
| Material | Leather, Silicone |
| Size | 10 |
| Thickness (Metric) | 1.1 to 1.3 mm |
| Liner Material | Aluminum/fleece |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?