Learn More
Scilabub™ Frosters™ Cryogenic Mid-Arm Glove
Product Code. p-8000440
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
Product Code. 11883831
Brand: Scilabub™ GLO/CMM
Only null left
Add to basket
Edge
Add to basket
Frosters™ Polyimide Cryogenic Gloves have been specially developed for the handling of very cold objects, typically for objects stored in the vapour phase of liquid nitrogen (ranging from -120°C to -190°C).
The middle layer is of special polyolefin/polyester micro-fibre, which is almost twice as effective in thermal insulation as a similar thickness of down. This minimises heat transfer without being so thick as to be cumbersome. The inner layer is of cotton for comfortable wear. They are made to EN 420 sizes and have been tested to EN388 (2, X, 2, X, ) and EN 511-contact cold 2, convective cold 2, Flexibility PASS. Avoid contact with alcohol-based products. To reduce bulk around the fingers, the stitch holes have not been sealed. In normal use this has not proved a problem, however, the gloves should not be immersed in liquid nitrogen and it is not advisable to use them above -80°C. Very durable, they retain their flexibility (albeit with some stiffening) for long periods in cold atmospheres. Polyimide outer material, polyolefin with some polyester as filling and cotton liner. Manufactured to EN511.Developed for handling very cold objects, typically those stored in the vapour phase of liquid nitrogen (in the range -120°C to -190°C).
Specifications
Color Blue
Material Fabric
Powdered Powder Free
Product Type Cryogenic Gloves
Size Medium
Sterility Non-sterile
Quantity 1 Pack
Resistant To Water