missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ White Cryobox with Dividers, 32 mm

Product Code. 15260554 Shop All Tenak Products
Click to view available options
Color:
White
Unit Size:
Pack of 36
This item is not returnable. View return policy

Product Code. 15260554

Brand: Tenak™ TE22225

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Cryobox with dividers for cryogenic storage.

  • Cryobox
  • For use with cryogenic storage applications

Specifications

Color White
Includes 9 x 9 Dividers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.