missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Cryo.s Round Bottom 4 mL Polypropylene Tube with External Thread Cap
GREENER_CHOICE

Product Code. p-7075345
Click to view available options
Closure Color:
Natural
Yellow
Includes:
ID Writing Area, 150 Inserts
Unit Size:
Pack of 300
This item is not returnable. View return policy

Product Code. 10563831

Brand: Greiner Bio-One undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.

  • With or without ID writing area
  • With Star foot base
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 100 white cap inserts
  • Polyethylener case
  • Dia. x Height: 12.4 x 83mm

Service Plan

No

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 12.4 mm
Product Type Cryogenic Storage Tube
Closure Color Natural
Capacity (Metric) 4 mL
Bottom Shape Round (Starfoot)
Material Polypropylene
Thread Style External
Includes ID Writing Area, 150 Inserts
Length (Metric) 83 mm
Packaging Polyethylene case
Sterility Sterile
Closure Type Threaded Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.