Learn More
Greiner Bio-One Cryo.s Freezing Tubes with Datamatrix Coding
GREENER_CHOICE
Product Code. 15674364
Change view
Click to view available options
Bottom Shape:
Conical
Thread Style:
Internal
Volume (Metric) Working:
1 mL
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bottom Shape Thread Style Volume (Metric) Working unitSize
15674364 Conical Internal 1 mL Pack of 500
15972282 Conical Internal 1 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15674364 Supplier Greiner Bio-One Supplier No. 1232632DG
Only null left
Add to basket
Add to basket
For storage of biological samples. Greiner Bio-One Cryo.s Freezing Tubes with Datamatrix Coding comes with colored screw caps for easy identification of biological samples.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic, endotoxin-free
  • High thermal resistance (196°C to +121°C)
  • Laser-written ECC200 Datamatrix coding
  • Strong resistance to mechanical and ch
Compliance
CE
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vial
Closure Color Natural
Bottom Shape Conical
Material Polyethylene
Thread Style Internal
Includes Cap inserts for labeling
Volume (Metric) Working 1 mL
Sterility Sterile
Closure Type Screw Cap