Learn More
Greiner Bio-One Cryo.s Cryogenic Storage Tube with Datamatrix Code
GREENER_CHOICE
Product Code. 15664364
Change view
Click to view available options
Capacity (Metric):
2 mL
Unit Size:
Pack of 500
Product Code. Capacity (Metric) unitSize
15664364 2 mL Pack of 500
1 options
Product Code. 15664364 Supplier Greiner Bio-One Supplier No. 1222632DG
Laser-written ECC200 Datamatrix Coding Service for cryogenic storage tubes.
  • Strong resistance to mechanical and chemical damage
  • High data capacity
  • Sequential sample IDs or consistent characters
  • Reed-Solomon error correction for high redundancy of encrypted information
  • Each Datamatrix is pro
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cryogenic Storage Tube
Description Datamatrix Code: 14 x 14, Sequence: G200000001, G200000002
Closure Color Natural
Capacity (Metric) 2 mL
Thread Style Internal
Includes Writing area
Closure Type Screw Cap