missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Gloves - Wrist Length

Product Code. 15649316 Shop All Thermo Scientific Products
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 15649316

Brand: Thermo Scientific™ 189574

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY

Specifications

Description Wrist Length
Color Blue
Size Medium
For Use With (Application) Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English) 12 in.
Length (Metric) 30.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.