missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™

Product Code. p-7224750
Click to view available options
Style:
Mid-Arm
Shoulder
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 11863881

Brand: Tempshield™ MAS

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

State of the art materials and multi-layered construction of Cryo-Gloves™ allows maximum thermal protection, flexibility and dexterity which are essential for function and safety. High performing, thermal inner lining wicks away moisture, maintaining comfort over extended periods.

  • Gives protection in ultra-cold environments down to -196°C
  • Maximum thermal protection, flexibility and dexterity
  • Inner lining wicks away moisture for added comfort during prolonged use
  • CE Cat. II, EN 511, EN 388, EN 420

Specifications

Certifications/Compliance (Europe) CE Category II, EN 511, EN 388, EN 420
Color Blue
Product Type Cryogenic Gloves
Size Small
Style Mid-Arm
Temperature (Metric) Max -196°C
For Use With (Application) Handling dry ice, low and ultra-low temperature freezers, closed cryogenic systems
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.