Learn More
ratiolab™ Cryo Boxes with Slip Lid, 8 x 8 Array
Product Code. 12831603
Change view
Click to view available options
Color:
Black
Blue
Red
Yellow
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12831603 Red Pack of 5
12811613 Black Pack of 5
12801613 Blue Pack of 5
12821603 Yellow Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12831603 Supplier ratiolab™ Supplier No. 5320212
Cryo boxes made of high-quality polypropylene. Available in natural and other colors.
These boxes are available in six modern colors and include corresponding vents for draining condensed water.
  • Boxes are adjustable in height
  • Autoclavable
  • Numerically coded
Specifications
Color Red
Material Polypropylene
Shape Square
Type Cryo Box
Tube Array 8 x 8
Withstand Temperature (Metric) -90°C to +121°C