Learn More
ratiolab™ Cryo Boxes with Slip Lid, 8 x 8 Array
Product Code. p-200106385
Click to view available options
Color:
Black
Blue
Red
Yellow
Unit Size:
Pack of 5
Product Code. 12811613
Brand: ratiolab™ 5320215
Only null left
Add to basket
Add to basket
Cryo boxes made of high-quality polypropylene. Available in natural and other colors.
These boxes are available in six modern colors and include corresponding vents for draining condensed water.
  • Boxes are adjustable in height
  • Autoclavable
  • Numerically coded
Specifications
Color Black
Material Polypropylene
Shape Square
Type Cryo Box
Tube Array 8 x 8
Withstand Temperature (Metric) -90°C to +121°C