missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYGS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557155
This item is not returnable.
View return policy
Description
Immunogen sequence: GTWAVYERPN FAGYMYILPQ GEYPEYQRWM GLNDRLSSCR AVHLPSGGQY KIQIFEKGDF SGQMYETTED CPSIMEQFHM REIHSCKVLE GVWIFYELP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. This gene encodes a protein initially considered to be a beta-crystallin but the encoded protein is monomeric and has greater sequence similarity to other gamma-crystallins. This gene encodes the most significant gamma-crystallin in adult eye lens tissue. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
Specifications
| CRYGS | |
| Polyclonal | |
| Unconjugated | |
| CRYGS | |
| AI327013; Beta-crystallin S; beta-crystallin S-like protein; CRYG8; CRYGS; crystallin gamma S; crystallin, gamma 8; crystallin, gamma polypeptide 8; crystallin, gamma S; CTRCT20; Gamma-crystallin S; gamma-crystallin S; beta-crystallin S; gamma-S-crystallin; gammaS-crystallin; opacity due to poor secondary fiber cell junction; Opj; recessive nuclear cataract; rncat; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1427, 689897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0C5E9, P22914 | |
| CRYGS | |
| Recombinant protein corresponding to Human CRYGS. Recombinant protein control fragment (Product #RP-96492). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction