missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYGS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569836
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 82%.
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. This gene encodes a protein initially considered to be a beta-crystallin but the encoded protein is monomeric and has greater sequence similarity to other gamma-crystallins. This gene encodes the most significant gamma-crystallin in adult eye lens tissue. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
Spezifikation
| CRYGS | |
| Polyclonal | |
| Unconjugated | |
| CRYGS | |
| AI327013; Beta-crystallin S; beta-crystallin S-like protein; CRYG8; CRYGS; crystallin gamma S; crystallin, gamma 8; crystallin, gamma polypeptide 8; crystallin, gamma S; CTRCT20; Gamma-crystallin S; gamma-crystallin S; beta-crystallin S; gamma-S-crystallin; gammaS-crystallin; opacity due to poor secondary fiber cell junction; Opj; recessive nuclear cataract; rncat; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 689897 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P0C5E9 | |
| CRYGS | |
| synthetic peptide directed towards the middle region of human CRYGS (aa 121-170). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur