missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYGN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21281-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| CRYGN | |
| Polyclonal | |
| Unconjugated | |
| CRYGN | |
| CRYGN, crystallin, gamma N, Gamma crystallin N, Gamma N crystallin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 155051, 214301 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VHL5, Q8WXF5 | |
| Crygn | |
| CRYGN Fusion Protein Ag15795 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction