missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYGD Monoclonal Antibody (2H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518655
This item is not returnable.
View return policy
Description
Peptide Sequence: SVRSCRLIPH SGSHRIRLYE REDYRGQMIE FTEDCSCLQD RFRFNEIHSL NVLEGSWVLY ELSNYRGRQY LLMPGDYRRY QDWGATNARV GSLRRVIDFS.
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. Four gamma-crystallin genes (gamma-A through gamma-D) and three pseudogenes (gamma-E, gamma-F, gamma-G) are tandemly organized in a genomic segment as a gene cluster. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
Specifications
| CRYGD | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P07320 | |
| CRYGD | |
| CRYGD (NP_008822, 75 a.a. ~ 174 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 2H3 | |
| Unconjugated | |
| CRYGD | |
| Aey4; Beta4abc; CACA; CCA3; CCP; Cryg-1; Cryg4; Crygd; cry-g-D; crystallin gamma D; crystallin, gamma D; Crystallin, gamma polypeptide 4; CTRCT4; DGcry-1; gamma crystallin 4; Gamma crystallin D; Gamma D crystallin; gamma-crystallin 1; Gamma-crystallin 2-2; gamma-crystallin 4; Gamma-crystallin D; gamma-D-crystallin; Len; lens opacity 12; Lop12; PCC | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1421 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction