missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CRYGC Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA527830

Product Code. 13411977

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A431, mouse eye. Predicted reactivity: Mouse (83%), Rat (85%), Dog (87%), Rhesus Monkey (96%), Chimpanzee (99%), Bovine (85%), Guinea pig (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. Four gamma-crystallin genes (gamma-A through gamma-D) and three pseudogenes (gamma-E, gamma-F, gamma-G) are tandemly organized in a genomic segment as a gene cluster. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRYGC
Polyclonal
Unconjugated
CRYGC
CCL; Cryg; Cryg3; Cryg-5; Crygc; crystallin gamma C; crystallin, gamma C; crystallin, gamma polypeptide 3; CTRCT2; Gammab-cry; Gamma-C-crystallin; gammaC-crystallin; gamma-crystallin 2-1; Gamma-crystallin 3; gamma-crystallin C; Gamma-crystallin IIIA; Len
Rabbit
Antigen affinity chromatography
RUO
12966, 1420
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P07315, Q61597
CRYGC
Recombinant protein encompassing a sequence within the center region of human gamma C Crystallin. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.