missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYGA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140030
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: CNSIRVDSGC WMLYERPNYQ GHQYFLRRGK Y Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.
CRYGA is a protein coding gene.
Specifications
| CRYGA | |
| Polyclonal | |
| Unconjugated | |
| CRYGA | |
| CRYG1; Cryg-4; CRYG5; CRYGA; CRY-g-A; CRY-gamma-A; crystallin gamma A; crystallin, gamma 1; crystallin, gamma A; crystallin, gamma polypeptide 1; DGcry-4; Gamma-A-crystallin; gammaA-crystallin; Gamma-crystallin 1-1; gamma-crystallin 4; Gamma-crystallin 5; gamma-crystallin A; Gamma-crystallin IVB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1418 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P11844 | |
| CRYGA | |
| Recombinant protein corresponding to Human CRYGA. Recombinant protein control fragment (Product #RP-108843). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction