missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYBB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | CRYBB3 |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695438
|
Novus Biologicals
NBP2-49196-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602307
|
Novus Biologicals
NBP2-49196 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRYBB3 Polyclonal antibody specifically detects CRYBB3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CRYBB3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS (pH 7.2), 40% Glycerol | |
| 1417 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Beta-B3 crystallin, beta-crystallin B3, CATCN2, CRYB3MGC125774, crystallin, beta B3, eye lens structural protein, MGC125772, MGC125773 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DAWSNSRDSDSLLSLRPLNIDSPHHKLHLFE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title