missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRYBA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567152
This item is not returnable.
View return policy
Description
Immunogen sequence: MTLQCTKSAGP WKMVVWDEDG FQG Highest antigen sequence identity to the following orthologs - mouse 88%, rat 83%.
Crystallins are the dominant structural components of the vertebrate eye lens.
Specifications
| CRYBA4 | |
| Polyclonal | |
| Unconjugated | |
| CRYBA4 | |
| beta crystallin A4 chain transcript PS; beta crystallin alpha 4 chain; beta-A4 crystallin; Beta-crystallin A4; Cryba4; crystallin beta A4; crystallin, beta A4; crystallin, beta polypeptide A4; CTA-445C9.1; CTRCT23; CYRBA4; eye lens structural protein; MCOPCT4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1413 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53673 | |
| CRYBA4 | |
| Recombinant Human CRYBA4. Recombinant protein control fragment (Product #RP-107797). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction