missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYBA4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92223-0.1ml
This item is not returnable.
View return policy
Description
CRYBA4 Polyclonal antibody specifically detects CRYBA4 in Human samples. It is validated for Western Blot
Specifications
| CRYBA4 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| Beta-A4 crystallin, beta-crystallin A4, crystallin, beta A4, crystallin, beta polypeptide A4, eye lens structural protein, MCOPCT4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-196 of human CRYBA4 (NP_001877.1). MTLQCTKSAGPWKMVVWDEDGFQGRRHEFTAECPSVLELGFETVRSLKVLSGAWVGFEHAGFQGQQYILERGEYPSWDAWGGNTAYPAERLTSFRPAACANHRDSRLTIFEQENFLGKKGELSDDYPSLQAMGWEGNEVGSFHVHSGAWVCSQFPGYRGFQYVLECDHHSGDYKHFREWGSHAPTFQVQSIRRIQQ | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1413 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction