missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYAB Polyclonal Antibody

CRYAB Polyclonal Antibody
Brand: Invitrogen PA518825
This item is not returnable.
View return policy
Description
CRYAB Polyclonal Antibody for Western Blot
Specifications
| CRYAB | |
| Polyclonal | |
| Unconjugated | |
| Cryab | |
| 2310001E10Rik; 3526402H21Rik; AACRYA; active transcription factor CREB; alpha B-crystallin; alpha crystallin B; alpha(B)-crystallin; alpha-B-crystallin; Alpha-crystallin B chain; alpha-crystallin B chain-like protein; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CMD1II; CREB; CREB1; CREB-1; Crya2; Crya-2; CRYAB; CryAB antibody; crystallin alpha B; crystallin, alpha 2; crystallin, alpha B; crystallin, alpha polypeptide 2; CTPP2; CTRCT16; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; epididymis secretory protein Li 101; heat shock protein beta-5; heat shock protein CryAB; heat shock protein HspB5; heat-shock 20 kD like-protein; HEL-S-101; HSPB5; HspB5 protein; MFM2; P23; Renal carcinoma antigen NY-REN-27; rosenthal fiber component; transactivator protein; Y protein | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 12955, 1410, 25420 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P02511, P23927, P23928 | |
| Cryab | |
| Synthetic peptide sequence (RLEKDRFSVNLD) corresponding to the internal amino acids of CRYAB (aa 69-80). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction