missing translation for 'onlineSavingsMsg'
Learn More

Cruzain-IN-1, MedChemExpress

Product Code. 16465146
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465146 100 mg 100mg
16454188 10 mg 10mg
16450537 50 mg 50mg
16446907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465146 Supplier Medchem Express Supplier No. HY10836

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cruzain-IN-1 is a covalent and reversible Cruzain inhibitor, with an IC50 of 10 nM.

Cruzain-IN-1 is a covalent and reversible Cruzain inhibitor, with an IC50 of 10 nM.

Specifications

Chemical Name or Material Cruzain-IN-1
CAS 1199523-24-4
Color White
Purity Grade Notes Research
Molecular Formula C14H10F2N6
Quantity 100 mg
Solubility Information DMSO : 80 mg/mL (266.43 mM; Need ultrasonic)
SMILES CCN1C2=NC(C#N)=NC(NC3=CC(F)=CC(F)=C3)=C2N=C1
Molecular Weight (g/mol) 300.27
Formula Weight 300.27
Percent Purity 99.9%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.