Learn More
Thermo Scientific Chemicals Crustacean Cardioactive Peptide
For cellular and molecular biology applications
Brand: Thermo Scientific™ J66453.#0
Code : NEW
Additional Details : Weight : 0.00005kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
-30°C to -10°C | |
957.10 | |
5 mg | |
Pro-Phe-Cys-Asn-Ala-Phe-Thr-Gly-Cys (Disulfide bridge: Cys3-Cys9) |
CCAP | |
Solid | |
Crustacean Cardioactive Peptide | |
RUO - research use only |
RUO – Research Use Only