missing translation for 'onlineSavingsMsg'
Learn More

CRTH-2/GPR44 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18409397 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409397 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409397 Supplier Novus Biologicals Supplier No. NBP176755AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CRTH-2/GPR44 Polyclonal antibody specifically detects CRTH-2/GPR44 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTH-2/GPR44
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human CRTH2. The immunogen is located within the first 50 amino acids of CRTH2. Amino Acid Squence: EQMSRLQSHSNTSIRYID
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 11251
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.