missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTH-2/GPR44 Antibody (54N3F8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£177.00 - £393.00
Specifications
| Antigen | CRTH-2/GPR44 |
|---|---|
| Clone | 54N3F8 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18119911
|
Novus Biologicals
NBP2-19014SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726733
|
Novus Biologicals
NBP2-19014 |
£393.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CRTH-2/GPR44 Monoclonal specifically detects CRTH-2/GPR44 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CRTH-2/GPR44 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| GPCR | |
| CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
| PTGDR2 | |
| IgG2b κ | |
| Protein G purified | |
| GPR44, or G-protein coupled receptor 44, is also designated as CRTH2 and CD294. GPR44 is expressed specifically on CD4+ Th2 cells and type 2 cytotoxic effector cells |
| 54N3F8 | |
| Western Blot 1-5 ug/ml, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 11251 | |
| A recombinant protein containing multiple external domains of GPR44 (CRTH2) was used as immunogen for the antibody. NP_004769 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title