missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTH-2/GPR44 Antibody (54N1H4), mFluor Violet 450 SE, Novus Biologicals™
Description
CRTH-2/GPR44 Monoclonal antibody specifically detects CRTH-2/GPR44 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CRTH-2/GPR44 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 54N1H4 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 |
| Host Species | Mouse |
| Immunogen | A recombinant protein containing multiple external domains of GPR44 (CRTH2) was used as immunogen for the antibody. NP_004769 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?