missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTC3 Polyclonal Antibody

CRTC3 Polyclonal Antibody
Brand: Invitrogen PA5109591
This item is not returnable.
View return policy
Description
CRTC3 Polyclonal Antibody for Western Blot
Specifications
| CRTC3 | |
| Polyclonal | |
| Unconjugated | |
| Crtc3 | |
| 2610312F20Rik; 6332415K15Rik; AI429792; BC011210; CREB regulated transcription coactivator 3; CREB-regulated transcription coactivator 3; Crtc3; RGD1309666; TORC3; TORC-3; transducer of CREB protein 3; transducer of regulated cAMP response element-binding protein (CREB) 3; transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64784 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.65 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6UUV7 | |
| Crtc3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human CRTC3 (NP_073606.3). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction