Learn More
CRTC2/TORC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12497-1-AP-150UL
 View more versions of this product
Product Code. 16840244
  • £349.00 / 150µL
Description
CRTC2, also known as TORC2 (Transducers Of Regulated cAMP Response Element-Binding (CREB)) 2 and the related protein CRTC1 are potent CREB coactivators that are exported from the nucleus in a CRM1-dependent manner via phosphorylation-dependent interactions. Studies suggest that their phosphorylation and nuclear/cytoplasmic shuttling play an important role in the regulation of gluconeogenesis by cAMP. CRTC2 is present in both B and T-lymphocytes and abundantly expressed in the thymus. Its activity is important in regulating the expression of genes involved in cellular energy metabolism while CRTC1 is essential for energy balance and fertility.
Specifications
Specifications
CRTC2/TORC2
Polyclonal
Unconjugated
CRTC2
CRTC2, RP11 422P24.6, TORC 2, TORC2, Transducer of CREB protein 2
Rabbit
Antigen Affinity Chromatography
RUO
200186, 310615, 74343
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3LRZ1, Q3U182, Q53ET0
Crtc2
CRTC2,TORC2 Fusion Protein Ag3167
150 μL
Primary
Human, Mouse, Rat