Learn More
CRTAM Antibody (Oxford Bio patent anti-CRTAM) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28400JF525
Product Code. 30171124
  • £328.00 / 0.10mL
Description
CRTAM Monoclonal antibody specifically detects CRTAM in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CRTAM
Monoclonal
Janelia Fluor 525
CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule
CRTAM/ CD355
0.1 mL
Immunology
56253
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Oxford Bio patent anti-CRTAM
50mM Sodium Borate
Human
Protein A purified
RUO
Primary
Human
Purified