Learn More
CRTAC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30251-1-AP-150UL
Description
The CRTAC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CRTAC1. This antibody recognizes human, mouse, rat antigen. The CRTAC1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CRTAC1 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| ASPIC, ASPIC1, cartilage acidic protein 1, CEP 68, CEP68, CRTAC1, FLJ10320 | |
| 150 μL | |
| 55118 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |