Learn More
CRTAC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30251-1-AP-20UL
 View more versions of this product
Product Code. 18065315
  • £135.00 / 20µL
Description
The CRTAC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CRTAC1. This antibody recognizes human, mouse, rat antigen. The CRTAC1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CRTAC1
600 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
ASPIC, ASPIC1, cartilage acidic protein 1, CEP 68, CEP68, CRTAC1, FLJ10320
20 μL
55118
Store at -20°C. Stable for one year after shipment.