missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRTAC1 Monoclonal Antibody (755315)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524362
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CRTAC1 encodes a glycosylated extracellular matrix protein that is found in the interterritorial matrix of articular deep zone cartilage. This protein is used as a marker to distinguish chondrocytes from osteoblasts and mesenchymal stem cells in culture. The presence of FG-GAP motifs and an RGD integrin-binding motif suggests that this protein may be involved in cell-cell or cell-matrix interactions. Copy number alterations in this gene have been observed in neurofibromatosis type 1-associated glomus tumors. Alternative splicing results in multiple transcript variants.
Specifications
| CRTAC1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9NQ79 | |
| CRTAC1 | |
| Mouse myeloma cell line NS0-derived recombinant human CRTAC1 Ser28-Cys661. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 755315 | |
| Unconjugated | |
| CRTAC1 | |
| 2810454P21Rik; 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; ASPIC1; AW047536; cartilage acidic protein 1; Cep68; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; Crtac1; Crtac1B; FLJ10320; Lotus; Protein CRTAC1-B; W307 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 55118 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction