missing translation for 'onlineSavingsMsg'
Learn More

CRT0066101 dihydrochloride, MedChemExpress

Product Code. 16499297
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499297

Brand: Medchem Express HY15698A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CRT0066101 dihydrochloride is a potent and specific PKD inhibitor with IC50 values of 1, 2.5 and 2 nM for PKD1, 2, and 3 respectively.

CRT0066101 dihydrochloride is a potent and specific PKD inhibitor with IC50 values of 1, 2.5 and 2 nM for PKD1, 2, and 3 respectively.

Specifications

Chemical Name or Material CRT0066101 dihydrochloride
CAS 1883545-60-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H24Cl2N6O
Quantity 5 mg
Solubility Information H2O : 50 mg/mL (121.56 mM; Need ultrasonic) ∣DMSO : 11.36 mg/mL (27.62 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1=CC=C(C2=CN(C)N=C2)C=C1C3=NC=CC(NC[C@H](N)CC)=N3.[H]Cl.[H]Cl
Molecular Weight (g/mol) 411.33
Formula Weight 411.33
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.