missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CRT 0066854 hydrochloride
GREENER_CHOICE

Product Code. 18763459
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18763459

Brand: Tocris Bioscience™ 5922/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PKCι and PKCζ inhibitor

CRT 0066854 hydrochloride is a protein kinase C (PKC) inhibitor. Inhibits PKCι and PKCζ (IC50 values are 132 and 639 nM, respectively). Restores polarized morphogenesis in oncogene expressing MDCK cells, decreases colony formation in HeLa cells and inhibits lethal giant larvae 2 (LLGL2) phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S)-3-Phenyl-N1-[5,6,7,8-tetrahydro-2-(4-pyridinyl)[1 ]benzothieno[2,3-d]pyrimidin-4-yl]-1,2-propanediamine dihydrochloride
CAS 2250019-91-9
Assay Percent Range >98%
Quantity 10 mg
Target Protein Kinase C Inhibitors
Molecular Formula C24H25N5S0.2 HCl
Formula Weight Observed MW: 488.48
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.