missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CRT 0066854 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CRT 0066854 hydrochloride is a protein kinase C (PKC) inhibitor. Inhibits PKCι and PKCζ (IC50 values are 132 and 639 nM, respectively). Restores polarized morphogenesis in oncogene expressing MDCK cells, decreases colony formation in HeLa cells and inhibits lethal giant larvae 2 (LLGL2) phosphorylation.
Specifications
Specifications
| Chemical Name or Material | (2 S)-3-Phenyl-N1-[5,6,7,8-tetrahydro-2-(4-pyridinyl)[1 ]benzothieno[2,3-d]pyrimidin-4-yl]-1,2-propanediamine dihydrochloride |
| CAS | 2250019-91-9 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Target | Protein Kinase C Inhibitors |
| Molecular Formula | C24H25N5S0.2 HCl |
| Formula Weight | Observed MW: 488.48 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction