missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crry/p65 Mouse anti-Rat, BUV615, Clone: 512, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 751636
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Crry/p65 Monoclonal antibody specifically detects Crry/p65 in Rat samples. It is validated for Flow Cytometry
Specifica
| Crry/p65 | |
| Monoclonal | |
| Brilliant Ultraviolet 615 | |
| Cr1; Crry; Cr1l | |
| Neuraminidase-treated rat peripheral blood erythrocytes | |
| RUO | |
| Rat | |
| IgG1 κ |
| Flow Cytometry | |
| 512 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto